missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcohol Dehydrogenase 1B Monoclonal Antibody (OTI5D7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525525
Additional Details : Weight : 0.01000kg
Description
The protein encoded by this gene is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This encoded protein, consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster.Specifications
Alcohol Dehydrogenase 1B | |
Monoclonal | |
0.69 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00325 | |
ADH1B | |
Full length human recombinant protein of ADH1B produced in HEK293T cell | |
100μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5D7 | |
Unconjugated | |
ADH1B | |
2700007F14Rik; acetaldehyde dehydrogenase 5; ADH, beta subunit; ADH1B; ADH2; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase subunit beta; aldehyde dehydrogenase 1 family member B1; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; aldehyde dehydrogenase X, mitochondrial; aldehyde reductase; ALDH class 2; Aldh1b1; ALDH5; Aldhx; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117 | |
Mouse | |
Affinity Chromatography | |
RUO | |
125 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |