Learn More
Steris™ Septihol™ Sterile Alcohol Solution
Septihol™ Sterile Alcohol Solution is gamma irradiated for sterility, and is a quick evaporating solution that is designed for use within cleanrooms and other critical environments to remove residue from hard surfaces.
Brand: Steris™ 6248Y4EC
Code : 04
Additional Details : CAS Number : 67-63-0 Weight : 1.00000kg
Description
- Ready to use sterile alcohol solution, eliminates errors in dilution preparation
- Gamma irradiated; SAL 10-6
- Removes organic and inorganic residue from hard, non-porous surfaces
- No stabilisers or denaturants - evaporates quickly
- Ideal for removal of process residues and for routine glove decontamination
- No rinsing
- Allows for high frequency use on most surfaces
- Reduces particulate levels
- Supports quality assurance and quality control
- Filtered to 0.22 μ
Specifications
Cleaning Solution | |
Sterile | |
Plastic Bottle | |
Filtered | |
Plastic bottle, fibreboard box |
Liquid | |
70% (v/v) USP Isopropyl Alcohol/30% (v/v) USP Purified Water | |
Sterile alcohol solution | |
Quick evaporating solution designed for use in cleanrooms and other critical environments to remove residue from hard surfaces | |
473 mL |