missing translation for 'onlineSavingsMsg'
Learn More

Alconox™ Liquinox™ Critical Cleaning Liquid Detergent

Product Code. p-4937103 Shop All Cole-Parmer Products
Click to view available options
Volume (English):
1 gal.
Container Type:
Case
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 11789679

Brand: Alconox™ 1777540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Concentrated, mild, anionic detergent for manual and ultrasonic cleaning

This concentrated, mild, anionic detergent is suitable for manual and ultrasonic cleaning.

Liquinox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, sodium xylene sulfonate, alkanolamide, and ethoxylated alcohol.

 

  • Concentrated
  • Phosphate free, biodegradable and readily disposable
  • Completely soluble in hard and soft water
  • Replaces corrosive acids and hazardous solvents
  • Dilute 1:100 (dilute 10mL in 1L of water)
  • Free rinsing—gives reliable results without interfering residues
  • Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants

 

Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors. Passes inhibitory residue test for water analysis. Used for phosphate sensitive analysis ware.

Used to remove:
Soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemicals and solvents.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life: 2 years from date of manufacture

Order Info

3.8L (1 gal.) makes 378.5L (100 gal.) cleaning solution.

Specifications

Product Type Critical Cleaning Liquid Detergent
Form Liquid
pH 8.5
Appearance Pale Yellow
Composition Anionic Detergent, Sodium Linear Alkylaryl Sulfonate, Sodium Xylene Sulfonate, Alkanolamide, Ethoxylated Alcohol, Water
Container Type Case
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Clean healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware
Volume (English) 1 gal.
Volume (Metric) 3.78 L
compliance-icons
Product Identifier
  • Liquinox
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains :
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.