missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 647 Azide, Triethylammonium Salt

Product Code. 10043964
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10043964

Brand: Molecular Probes™ A10277

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The far red-fluorescent Alexa Fluor™ 647 azide is reactive with terminal alkynes via a copper-catalyzed click reaction.

The far red-fluorescent Alexa Fluor™ 647 azide is reactive with terminal alkynes via a copper-catalyzed click reaction. The bright and photostable fluorophore can be for used with flow cytometry, microscopy, and HCS

Dissolve in high-quality anhydrous DMSO for stock concentrations of 1 to 10 mM.

TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 0.5 mg
Product Type Azide
Molecular Weight (g/mol) ∼850 g/mol
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate, and protect from light.
Chemical Reactivity Alkyne
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Amine, Azide
Label or Dye Alexa Fluor™ 647
Label Type Alexa Fluor
Color Far-red
Product Line Alexa Fluor, Molecular Probes
Emission 665 nm
Excitation 650 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.