missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alexa Fluor™ 647-DYKDDDDK Peptide
Alexa Fluor™ 647-DYKDDDDK Peptide
Brand: Invitrogen™ PV6036
Code : 50
Additional Details : Weight : 0.50000kg
Description
A control reagent for use with the Eu-anti-DYKDDDDK Antibody (PV6026 and PV6027).Specifications
Alexa Fluor™ | |
400 nM Alexa Fluor 647-DYKDDDDK Peptide in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl, pH 7.5 (HEPES buffered saline). Store at -20°C and protect from light. | |
Alexa Fluor™ 647 | |
0.4 mL | |
Dry Ice |
Far-Red | |
Frozen | |
Alexa Fluor Dyes | |
Drug Discovery, Immunology |
For Research Use Only. Not for use in diagnostic procedures.