missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Alexa Fluor™ 647 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed
Alexa Fluor™ 647 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed
Brand: Molecular Probes™ A31634
Code : 50
Additional Details : Weight : 0.00454kg
Description
Alexa Fluor™ SFX Kits contain Image-iT™ FX signal enhancer (Cat. no. I36933) plus one of sixteen different Alexa Fluor™ dye-labeled secondary antibodies. These kits provide 400μg (0.2 mL of 2 mg/mL) of either goat anti-mouse IgG or goat anti-rabbit IgG antibody (as a standard or highly cross-adsorbed preparation) conjugated to Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594, or Alexa Fluor™ 647 dye, four of our most commonly used Alexa Fluor™ dyes. The Alexa Fluor™ 647 goat anti-rabbit IgG, highly cross-adsorbed, included with this kit is also available in a 1000μg unit size (Cat. no. A21245).Specifications
Alexa Fluor 647 | |
Goat | |
RUO, RUO | |
Secondary | |
Rabbit | |
Rabbit | |
Alexa Fluor™, Molecular Probes™ |
Alexa Fluor™ 647 Goat Anti-Rabbit SFX Kit, 1kit, Goat Host, Rabbit Species Reactivity, Alexa Fluor™ 647 Conjugate, Isotype, 2 to 8°C Storage Temperature, Room Temperature Shipping Condition | |
1 Kit | |
Whole Antibody | |
1 Each | |
Store in refrigerator (2–8°C) and protect from light. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.