missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlgiMatrix™ 24-Well, 3D Culture System, Flat-Bottom Microplate

Catalog Number 10751574
Click to view available options
1/Pk.
4/Pk.
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Color: Clear; Sterility: Sterile; Lid: With Lid

The Algimatrix™ 3D Culture System provides a three-dimensional matrix that has the potential to create cell culture conditions that more closely reflect normal cell morphology and behavior. It has relevance for both academic and industrial researchers who want to culture hepatocytes and other spheriod forming cells. Algimatrix™ alginate sponge is provided in a 24 well format that fits well into the workflow of both lower and higher-throughput users. The larger well size enables growth of more cells for downstream applications such as transfection.
TRUSTED_SUSTAINABILITY
Lid With Lid
Content And Storage Higher-fidelity, animal-free scaffold for 3D cell culture that mimics in vivo conditions. More consistent results than animal-derived matrices. Ready to use, stored at room temperature and stable up to 18 months. Protocols available.
Packaging 1 Pack
Product Line Algimatrix, Gibco
Color Clear
Product Type Microplate
No. of Wells 24
Sterility Sterile
Surface Treatment AlgiMatrix 3D Culture System
Well Shape Flat Bottom (F-well)
Purity or Quality Grade Cell Culture Grade
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'