missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Alginic acid, sodium salt
£23.80 - £305.00
Chemical Identifiers
CAS | 9005-38-3 |
---|---|
Molecular Formula | (C6H7O7)A(C6H7O7)BNa |
MDL Number | MFCD00081310 |
Synonym | Algin, Sodium alginate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521631
|
Thermo Scientific Chemicals
177770050 |
5 g | Glass bottle |
£23.80
5g |
In Stock
Log in to see stock. |
||||
10062400
|
Thermo Scientific Chemicals
177772500 |
250 g | Plastic Bottle |
£50.30
250g |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||||
10215710
|
Thermo Scientific Chemicals
177775000 |
500 g | Plastic Bottle |
£76.00
500g |
In Stock
Log in to see stock. |
||||
11936991
|
Thermo Scientific Chemicals
177770025 |
2.5 kg | Plastic Bottle |
£305.00
2.50kg |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9005-38-3 | |
(C6H7O7)A(C6H7O7)BNa | |
MFCD00081310 | |
Algin, Sodium alginate |
Specifications
9005-38-3 | |
MFCD00081310 | |
Alginic acid, sodium salt |
(C6H7O7)A(C6H7O7)BNa | |
Algin, Sodium alginate |
Safety and Handling
missing translation for 'rtecsNumber' : AZ5820000
missing translation for 'tsca' : TSCA
RUO – Research Use Only