Learn More
Thermo Scientific™ aLICator LIC Cloning and Expression Kit 2 (N-terminal His-tag/EK)
Get fast and efficient ligation-independent cloning and tight regulation of gene expression in E. coli. with this cloning and expression system.
Brand:  Thermo Scientific™ K1251
Product Code. 10198802
  • £248.00 / 20 reactions
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
aLICator LIC Cloning and Expression System is designed for fast and efficient ligation independent cloning and tight regulation of gene expression in E. coli. The pLATE bacterial expression vectors are designed for high levels of target protein expression in concert with minimal background (uninduced) expression, which permits expression of proteins that are toxic to E. coli cells. To streamline and facilitate the process of insert cloning into the expression vector, the aLICator system uses directional LIC cloning technology, a rapid procedure that provides high-cloning efficiencies.
The tightly regulated expression and fast, efficient directional cloning makes the aLICator LIC Cloning and Expression System the best choice for routine and toxic gene cloning and expression in E. coli.
Highlights
  • High efficiency LIC cloning
  • Tight control of gene expression
  • High yield expression
  • Versatile -tagged or untaged protein expression with tag removal option
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ampicillin (AmpR)
Kit
PCR Cloning
LIC Cloning and Expression Kit
His Tag (6x)
pLATE
LIC Cloning
PCR Products
aLICator
T7
20 Reactions