missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Aliphatic Amine Latex Beads, 2% w/v, 2μm

Aliphatic Amine Latex Beads, 2% w/v, 2 μm

Brand:  Molecular Probes™ A37366

Code : 50

Additional Details : Weight : 0.00525kg

Product Code. 11594087

  • £271.00 / 15mL
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This aliphatic amine latex has a high density of amine groups attached to the terminus of an aliphatic six-carbon spacer arm. This product can be used to covalently couple proteins. Location of the amine at the end of the spacer arm minimizes difficulties with steric hindrance and conformation since the group has a greater freedom to rotate. In addition, spacer arms improve the kinetics of latex agglutination reactions. The particles are stabilized by the positively-charged amine groups under low to neutral pH conditions. Care should be taken not to use them under high pH.

Microspheres Are Available in Many Sizes and Surface Functionalities
  • 25 sizes to choose from—0.02 microns to 16 microns
  • 20 different surface—for ready conjugation to virtually any material
  • Surfactant free—ultraclean
  • Various quantities—from milliliter to 500-liter amounts

    Key Applications of Microspheres
  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

    Get More Information
    Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

    We’ll Make a Custom Microsphere Product for You
    If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • Specifications

    Specifications

    0.02w/v
    Aliphatic Amine Latex Beads, 15mL, 2% w/v Concentration, 2μm Particle Size, White, Suspension Form, De-ionized Water Medium, Super Active Latex Beads, Room Temperature, 2 to 8°C Storage Temperature, Ideal Materials for the Adsorption of Materials S
    Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents)
    Suspension
    17 A2/NH2 (Amino Group), 97 A2/COOH (Carboxyl Group)
    15 mL
    Aliphatic Amine
    2 μm
    2°C to 8°C
    White
    Polystyrene
    1 Each
    1.055g/cm3 at 20°C
    1.591 at 20°C
    15 mL
    2 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.