Learn More
Aliquat∣r 336 TG, Thermo Scientific Chemicals
£23.60 - £271.00
Chemical Identifiers
CAS | 63393-96-4 |
---|---|
Molecular Formula | C25H54ClN |
MDL Number | MFCD00011862 |
InChI Key | XKBGEWXEAPTVCK-UHFFFAOYSA-M |
Synonym | methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride |
PubChem CID | 21218 |
ChEBI | CHEBI:75286 |
IUPAC Name | methyl(trioctyl)azanium;chloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433477
|
Thermo Scientific Alfa Aesar
|
100 mL |
£23.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443477
|
Thermo Scientific Alfa Aesar
|
500 mL |
£78.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453477
|
Thermo Scientific Alfa Aesar
A17247.0F |
2500 mL |
£271.00
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.
Solubility
Miscible with benzene and chloroform.
Notes
Incompatible with strong oxidizing agents, acids and bases.
Chemical Identifiers
63393-96-4 | |
MFCD00011862 | |
methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride | |
CHEBI:75286 |
C25H54ClN | |
XKBGEWXEAPTVCK-UHFFFAOYSA-M | |
21218 | |
methyl(trioctyl)azanium;chloride |
Specifications
63393-96-4 | |
>240°C | |
C25H54ClN | |
MFCD00011862 | |
Hygroscopic | |
Miscible with benzene and chloroform. | |
CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-] | |
404.164 | |
CHEBI:75286 | |
Aliquat 336 |
0.88 | |
132°C (269°F) | |
1.4465 | |
UN2810 | |
methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride | |
XKBGEWXEAPTVCK-UHFFFAOYSA-M | |
methyl(trioctyl)azanium;chloride | |
21218 | |
404.17 |
Safety and Handling
GHS H Statement
H301-H318-H315
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335-H360FD-H373
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 264-120-7
missing translation for 'rtecsNumber' : BR8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Aliquat 336 TG
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Quaternary ammoniumcompounds, tri-C8- 10-alkylmethyl, chlorides, 1-Octanol, 1-Decanol, Amines, tri-
RUO – Research Use Only