missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Alizarin, 97%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 287.0°C to 289.0°C |
| Boiling Point | 430.0°C |
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Dye Content | 96.0% min. |
| Assay Percent Range | 97% |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in aromatic solvents, ether and hot, methanol, moderately soluble in ethanol |
| Infrared Spectrum | Authentic |
| Formula Weight | 240.21 |
| Loss on Drying | 2% max. |
| Show More |
Seguridad y manipulación
Nombre del producto
- Alizarin
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido