missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Alizarin Red S, pure, certified
£81.60 - £234.00
Chemical Identifiers
CAS | 130-22-3 |
---|---|
Molecular Formula | C14H7NaO7S |
Molecular Weight (g/mol) | 342.253 |
MDL Number | MFCD00013049 |
InChI Key | HFVAFDPGUJEFBQ-UHFFFAOYSA-M |
Synonym | 9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate |
PubChem CID | 3955344 |
ChEBI | CHEBI:87358 |
IUPAC Name | sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate |
SMILES | C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10381101
|
Thermo Scientific Chemicals
400480250 |
25 g |
£81.60
25g |
In Stock
Log in to see stock. |
|||||
15234724
|
Thermo Scientific Chemicals
400481000 |
100 g |
£234.00
100g |
In Stock
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
130-22-3 | |
342.253 | |
HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
3955344 | |
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate |
C14H7NaO7S | |
MFCD00013049 | |
9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate | |
CHEBI:87358 | |
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] |
Specifications
130-22-3 | |
C14H7NaO7S | |
9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate | |
HFVAFDPGUJEFBQ-UHFFFAOYSA-M | |
C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+] | |
342.253 | |
CHEBI:87358 | |
342.26 | |
Glass bottle | |
Red | |
Alizarin Red S |
0.97 to 1.07 (for the first maximum) (P-15/P+15) | |
MFCD00013049 | |
Solubility in water: moderately soluble. Other solubilities: soluble in ethanol | |
Authentic | |
sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate | |
3955344 | |
554 to 558nm 590 to 599nm | |
Pure | |
from pH 3.5 (yellow) to pH 6.5 (pink) and from pH 9.4 (orange-brown) to pH 12.0 (violet) | |
Powder |
Safety and Handling
Warning
missing translation for 'einecsNumber' : 204-981-8
missing translation for 'rtecsNumber' : CB1095300
missing translation for 'tsca' : TSCA
Product Identifier
- Alizarin Red S
RUO – Research Use Only