missing translation for 'onlineSavingsMsg'
Learn More

Alizarin Yellow R sodium salt, Thermo Scientific Chemicals

Product Code. 11358977
10 g
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1718-34-9
Molecular Formula C13H8N3NaO5
Molecular Weight (g/mol) 309.213
MDL Number MFCD00067121
InChI Key HKSANXZSKDUDBD-UHFFFAOYSA-M
Synonym alizarin yellow r sodium salt, acmc-1bqov, alizarin yellow r, sodium salt,
PubChem CID 44135758
IUPAC Name sodium;3-[(4-nitrophenyl)hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

Color Brown to Yellow
Physical Form Powder
Quantity 10 g
Beilstein 962365
Formula Weight 309.22
Odor Odorless
Appearance Dark yellow to brown powder
Chemical Name or Material Alizarin Yellow R sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alizarin Yellow R sodium salt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.