missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ALK Split FISH Probe

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

£1270.00 - £2085.00

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 2p23, 2p23
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16063395
View Documents Promotion Details
Abnova™
FS0001.200uL
200 μL
£2085.00
200µL
Please to purchase this item. Need a web account? Register with us today!
16053395
View Documents Promotion Details
Abnova™
FS0001.100uL
100 μL
£1270.00
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Specifications

Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
2p23, 2p23
ALK Split FISH Probe
FITC, Texas Red
DAPI Counterstain (1500 ng/mL ) 250 uL
ALK, ALK
Videos
SDS
Documents

Documents

Product Certifications