missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ALK Split FISH Probe
Abnova FISH probes are ensured to provide researchers an effective tool to confirm various genetic amplification, rearrangements, deletions, and aneuploidy.
Brand: Abnova™ FS0047.200uL
This item is not returnable.
View return policy
Description
- Gene name: ALK
- Gene Alias: CD246, Ki-1, TFG/ALK
- Entrez GeneID: 238
- Gene Description: anaplastic lymphoma receptor tyrosine kinase
Specifications
Store at 4°C in the dark. | |
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. | |
ALK Split FISH Probe | |
200 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ ALK Split FISH Probe > Quantity: 200μL;
For research use only
Spot an opportunity for improvement?Share a Content Correction