missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Alkali-Cation™ Yeast Transformation Kit

Provides all of the reagents required to perform a quick and easy transformation. Alkali-Cation™ Yeast Transformation Kit is easy to use, robust, and its scalability makes it great for the high-efficiency transformation needed in two hours.

Brand:  MP Biomedicals™ 112200200

Product Code. 11343159

  • £534.00 / 250 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • An efficient, non-electrical method for transforming yeast with linear or plasmid DNA
  • Host cells are made competent by treatment with a Lithium/Cesium Acetate mixture
  • The entire procedure may be completed in less than 90 minutes
  • Routinely provides a transformation efficiency of greater than 104 transformants per μg of DNA
  • Has been successfully used to transform linear DNA into Pichia pastoris for protein expression studies
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S. cerevisiae, S. pombe, S. albicans, many strains
Kit
Yeast transfomation
500 μL or 3 to 4 mm colony
4°C
Yeast Culture
250 Preps
High Efficiency Yeast Transformation Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.