Learn More
Linden™ Alkaline Button Batteries
Providing long shelf life for general laboratory purposes. Linden™ Alkaline Button Batteries have a leak-resistant construction.
£12.31 - £12.35
Specifications
Product Type Alkaline Battery
Quantity 1 Ea.
Voltage 1.5 V
View More Specs
Products 2
Product Code Brand Amperage Price Quantity & Availability  
Product Code Brand Amperage Price Quantity & Availability  
12717565
View Documents
Linden™
H-GP-186-CD10
70 mAh capacity
£12.31
Each
In Stock
to see stock.
Edge
Add to basket
12737565
View Documents
Linden™
H-GP-189-CD10
44 mAh capacity N/A N/A N/A
Description
Description
  • Available in a variety of different voltages
  • In button form
  • High energy density
Specifications
Specifications
Alkaline Battery
1 Ea.
1.5 V
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Linden™ Alkaline Button Batteries