Learn More
Brady™ All-weather Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170822
Additional Details : Weight : 0.12900kg
 View more versions of this product
Product Code. 17970700
  • £33.87 / Each
Estimated Shipment: 13-08-2024
to see stock.
Description
Description
  • Ideal for labeling pipes, equipment, shelves and other industrial surfaces
  • Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
  • Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
  • Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White, Clear
Continuous
Top
Labels
Component and Equipment Labeling, Electrical Labeling, Facility Identification, Floor Marking, Laboratory Labeling, Lean and 5S Labeling, Pipe and Valve Marking, Warehouse Marking
1 Tape/Cartridge
0.004 in.
20 ft.
1.3 cm
RoHS Compliant to 2011/65/EU (Pass 2015/863)
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
BMP41, BMP51, M511
-40°F to +180°F
20 ft. x 0.5 in.
0.5 in.
0.5 in.