missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, 98%, Thermo Scientific Chemicals

Product Code. 11403047
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allantoin is a useful biomarker for oxidative stress. It is a reactant for Synthesis of sulofenur analogs for use as antitumor agents. It is used as a cosmetic agent.

Solubility
Slightly soluble in water. Freely soluble in alkalis

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.117
MDL Number MFCD00005260
InChI Key POJWUDADGALRAB-UHFFFAOYSA-N
Synonym allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
PubChem CID 204
ChEBI CHEBI:15676
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES C1(C(=O)NC(=O)N1)NC(=O)N

Specifications

Melting Point ∼230°C (decomposition)
Quantity 250 g
Beilstein 102364
Merck Index 14,258
Solubility Information Slightly soluble in water. Freely soluble in alkalis
Formula Weight 158.12
Percent Purity 98%
Chemical Name or Material Allantoin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'