Learn More
Allihn Condenser with Ground Cone and Socket
Ideal for reflux applications.
£248.00 - £285.73
Product Code | Brand | Standard Taper Joint No | Length (Metric) Jacket | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Length (Metric) Jacket | Price | Quantity & Availability | ||||
11381845
|
Quickfit
CX7/33/SC |
24/29 | 545 mm |
£248.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15507790
|
Quickfit
CX7/44/SC |
29/32 | 550 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Borosilicate Glass | |
Allihn Condenser |