Learn More
Chemical Identifiers
CAS | 107-18-6 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00002920 |
InChI Key | XXROGKLTLUQVRX-UHFFFAOYSA-N |
Synonym | allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench |
PubChem CID | 7858 |
ChEBI | CHEBI:16605 |
IUPAC Name | prop-2-en-1-ol |
SMILES | OCC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11401960
|
Thermo Scientific Alfa Aesar
A15026.AE |
100 mL |
£27.03
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11670212
|
Thermo Scientific Alfa Aesar
A15026.AP |
500 mL |
£44.82
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15413107
|
Thermo Scientific Alfa Aesar
A15026.AU |
1 L |
£69.75
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, acids, oleum and alkali metals. Polymerizes during prolonged storage.
Chemical Identifiers
107-18-6 | |
58.08 | |
XXROGKLTLUQVRX-UHFFFAOYSA-N | |
7858 | |
prop-2-en-1-ol |
C3H6O | |
MFCD00002920 | |
allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench | |
CHEBI:16605 | |
OCC=C |
Specifications
107-18-6 | |
0.852 | |
22°C (71°F) | |
Pungent | |
CH2=CHCH2OH | |
MFCD00002920 | |
605307 | |
allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench | |
XXROGKLTLUQVRX-UHFFFAOYSA-N | |
prop-2-en-1-ol | |
7858 | |
58.08 | |
Liquid |
-129°C | |
95°C to 97°C | |
≥98% | |
C3H6O | |
1.413 | |
UN1098 | |
14,286 | |
Miscible with water. | |
OCC=C | |
58.08 | |
CHEBI:16605 | |
≥98% | |
Allyl alcohol |
Safety and Handling
GHS H Statement
H225-H301-H311-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H301-H310+H330-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALLYL ALCOHOL
missing translation for 'einecsNumber' : 203-470-7
missing translation for 'rtecsNumber' : BA5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Allyl alcohol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Allyl alcohol
RUO – Research Use Only