Learn More
Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide, Thermo Scientific Chemicals
£30.45 - £94.87
Chemical Identifiers
CAS | 106-95-6 |
---|---|
Molecular Formula | C3H5Br |
Molecular Weight (g/mol) | 120.977 |
MDL Number | MFCD00000244 |
InChI Key | BHELZAPQIKSEDF-UHFFFAOYSA-N |
Synonym | allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide |
PubChem CID | 7841 |
IUPAC Name | 3-bromoprop-1-ene |
SMILES | C=CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429333
|
Thermo Scientific Alfa Aesar
A11766.30 |
250 g |
£30.45
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419333
|
Thermo Scientific Alfa Aesar
A11766.0B |
1000 g |
£94.87
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.
Solubility
Slightly miscible with water.
Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkali metals, alkaline earth metals, amides, amines and powdered metals.
Chemical Identifiers
106-95-6 | |
120.977 | |
BHELZAPQIKSEDF-UHFFFAOYSA-N | |
7841 | |
C=CCBr |
C3H5Br | |
MFCD00000244 | |
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide | |
3-bromoprop-1-ene |
Specifications
106-95-6 | |
1.398 | |
-2°C (28°F) | |
C3H5Br | |
1.47 | |
UN1099 | |
14,288 | |
Slightly miscible with water. | |
C=CCBr | |
120.977 | |
120.98 | |
Liquid |
°C to 119°C | |
70°C to 71°C | |
99% | |
CH2=CHCH2Br | |
MFCD00000244 | |
Light sensitive | |
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide | |
BHELZAPQIKSEDF-UHFFFAOYSA-N | |
3-bromoprop-1-ene | |
7841 | |
99% | |
Allyl Bromide, Stabilized with 300 to 1000ppm Propylene oxide |
Safety and Handling
GHS H Statement
H225-H301-H331-H340-H350-H314
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H331-H314-H335-H340-H350
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ALLYL BROMIDE
missing translation for 'einecsNumber' : 203-446-6
missing translation for 'rtecsNumber' : UC7090000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Allyl bromide, stabilized with 300-1000ppm Propylene oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Allyl bromide, Propylene oxide
RUO – Research Use Only