missing translation for 'onlineSavingsMsg'
Learn More

Allyl bromide, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10609283
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
5 L
10 L
Packaging:
Glass bottle
Metal Can
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-95-6, 75-56-9
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00000244
InChI Key BHELZAPQIKSEDF-UHFFFAOYSA-N
Synonym allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
PubChem CID 7841
IUPAC Name 3-bromoprop-1-ene
SMILES C=CCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -119.0°C
Color Colorless
Density 1.3900g/mL
Boiling Point 70.0°C to 71.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Odor The liquid has a pungent odor
Packaging Plastic drum
Linear Formula H2C=CHCH2Br
Refractive Index 1.4680 to 1.4700
Quantity 10 L
Beilstein 01,201
Fieser 13,219
Merck Index 15,279
Specific Gravity 1.39
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol, chloroform, ether,, carbontetrachloride and carbon disulfide
Stabilizer 300ppm propylene oxide
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl bromide, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl bromide, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Allyl bromide, Propylene oxide
  • REA17-CMR1-Restricted to professional users

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'