missing translation for 'onlineSavingsMsg'
Learn More

Allyl bromide, 99%, stabilized

Product Code. 10706131 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
5 L
10 L
Packaging:
Glass bottle
Metal Can
Plastic drum
Unit Size:
10 L
2.5 L
250mL
5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10706131

Brand: Thermo Scientific Acros 102902500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 106-95-6 | C3H5Br | 120.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-95-6, 75-56-9
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00000244
InChI Key BHELZAPQIKSEDF-UHFFFAOYSA-N
Synonym allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
PubChem CID 7841
IUPAC Name 3-bromoprop-1-ene
SMILES C=CCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -119.0°C
Color Colorless
Density 1.3900g/mL
Boiling Point 70.0°C to 71.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Odor The liquid has a pungent odor
Packaging Glass bottle
Linear Formula H2C=CHCH2Br
Refractive Index 1.4680 to 1.4700
Quantity 250 mL
Beilstein 01,201
Fieser 13,219
Merck Index 15,279
Specific Gravity 1.39
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol, chloroform, ether,, carbontetrachloride and carbon disulfide
Stabilizer 300ppm propylene oxide
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl bromide, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl bromide, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Allyl bromide, Propylene oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.