missing translation for 'onlineSavingsMsg'
Learn More

Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals

Product Code. 10572741 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10572741

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-23-5
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00004894
InChI Key ULIKDJVNUXNQHS-UHFFFAOYSA-N
Synonym allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol
PubChem CID 13367
IUPAC Name prop-2-ene-1-thiol
SMILES SCC=C

Specifications

Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 67°C to 68°C
Flash Point 18°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula H2C=CHCH2SH
Refractive Index 1.4745 to 1.4785
Quantity 25 g
Beilstein 01, 440
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 74.13
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material Allyl mercaptan
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allyl mercaptan, 70%, remainder mainly sulphide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Propene-1-thiol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.