missing translation for 'onlineSavingsMsg'
Learn More

Allyl methyl sulfide, 98%

Product Code. p-7026492
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11490697

Brand: Thermo Scientific Alfa Aesar B22514.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10152-76-8 | C4H8S | 88.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10152-76-8
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00008657
InChI Key NVLPQIPTCCLBEU-UHFFFAOYSA-N
Synonym allyl methyl sulfide, 1-propene, 3-methylthio, allyl methyl sulphide, 3-methylthio-1-propene, sulfide, allyl methyl, methyl propenyl sulfide, unii-v7qi1r316c, methyl allyl sulfide, ccris 7066
PubChem CID 66282
IUPAC Name 3-methylsulfanylprop-1-ene
SMILES CSCC=C

Specifications

Density 0.803
Boiling Point 91°C to 93°C
Flash Point 18°C (65°F)
Odor Stench
Refractive Index 1.4714
Quantity 5 g
UN Number UN1993
Formula Weight 88.17
Percent Purity 98%
Chemical Name or Material Allyl methyl sulfide
compliance-icons
Product Identifier
  • Allyl methyl sulphide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Allyl methyl sulphide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.