missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Product Code. 11331707
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5296-64-0
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00014957
InChI Key QGNRLAFFKKBSIM-UHFFFAOYSA-N
Synonym allyl phenyl sulfide, allylthio benzene, allyl phenyl sulfane, benzene, 2-propenylthio, allylphenyl sulfide, phenyl allyl sulfide, allylphenylthioether, allyl phenyl ;sulfane, sulfide, allyl phenyl
PubChem CID 79180
IUPAC Name prop-2-enylsulfanylbenzene
SMILES C=CCSC1=CC=CC=C1

Specifications

Density 1.024
Boiling Point 223°C to 225°C
Flash Point >98°C (208°F)
Odor Unpleasant
Refractive Index 1.574
Quantity 50 g
Beilstein 1856940
Sensitivity Air Sensitive
Formula Weight 150.24
Percent Purity 97%
Chemical Name or Material Allyl phenyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'