missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Color | White |
Density | 0.7630g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | H2C=CHCH2NH2 |
Refractive Index | 1.4200 to 1.4230 (20°C, 589nm) |
Quantity | 1 L |
Show More |
Safety and Handling
Product Identifier
- Allylamine
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- H310-Fatal in contact with skin.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'