missing translation for 'onlineSavingsMsg'
Learn More
Allylamine, 98%, Thermo Scientific Chemicals Product Code.: 10637202

Allylamine, 98%, Thermo Scientific Chemicals

Product Code. 10637202
50 mL, Glass bottle
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10637202

Brand: Thermo Scientific Acros 220730500

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-11-9
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.10
MDL Number MFCD00008199
InChI Key VVJKKWFAADXIJK-UHFFFAOYSA-N
Synonym allylamine, 2-propen-1-amine, monoallylamine, 3-aminopropylene, 3-aminopropene, allyl amine, 2-propenamine, 3-amino-1-propene, 2-propenylamine, polyallylamineShow MoreShow Less
PubChem CID 7853
IUPAC Name prop-2-en-1-amine
SMILES NCC=C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -88°C
Color White
Density 0.7630g/mL
Boiling Point 53°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2NH2
Refractive Index 1.4205 to 1.4225
Quantity 50 mL
Beilstein 04,205
Merck Index 15,278
Specific Gravity 0.763
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether
Viscosity 0.44 mPa.s (25°C)
Formula Weight 57.09
Percent Purity 98%
Grade Extra Pure
Physical Form Crystalline or Granular Powder
Chemical Name or Material Allylamine, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398546 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0409035 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0414858 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberA0409036 Certificate TypeCertificate of Analysis Date07/06/2020 Catalog Number
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allylamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.