missing translation for 'onlineSavingsMsg'
Learn More

Allylchlorodimethylsilane, 94%, Thermo Scientific Chemicals

£38.99 - £135.68

Chemical Identifiers

CAS 4028-23-3
Molecular Formula C5H11ClSi
Molecular Weight (g/mol) 134.678
MDL Number MFCD00000506
InChI Key KMVZWUQHMJAWSY-UHFFFAOYSA-N
Synonym allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7
PubChem CID 77646
IUPAC Name chloro-dimethyl-prop-2-enylsilane
SMILES C[Si](C)(CC=C)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11415886
View Documents
Thermo Scientific Alfa Aesar
B22237.06
5 g
£38.99
5g
Please to purchase this item. Need a web account? Register with us today!
11374609
View Documents
Thermo Scientific Alfa Aesar
B22237.09
10 g
£61.91
10g
Please to purchase this item. Need a web account? Register with us today!
11425886
View Documents
Thermo Scientific Alfa Aesar
B22237.14
25 g
£135.68
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1?,4,4?-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1′,4,4′-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

Solubility
Soluble in most organic solvents; reacts with alcohols, amines, and water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.
Specifications

Chemical Identifiers

4028-23-3
134.678
KMVZWUQHMJAWSY-UHFFFAOYSA-N
77646
C[Si](C)(CC=C)Cl
C5H11ClSi
MFCD00000506
allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7
chloro-dimethyl-prop-2-enylsilane

Specifications

110°C to 112°C
C5H11ClSi
UN2985
5°C (41°F)
Soluble in most organic solvents; reacts with alcohols,amines,and water.
C[Si](C)(CC=C)Cl
134.678
134.68
1.429
0.896
4028-23-3
MFCD00000506
1737978
allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7
KMVZWUQHMJAWSY-UHFFFAOYSA-N
chloro-dimethyl-prop-2-enylsilane
77646
97%
Moisture sensitive
Allylchlorodimethylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 223-706-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only