missing translation for 'onlineSavingsMsg'
Learn More

Allylmagnesium chloride, 1.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10464643
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-05-1, 592-42-7
Molecular Formula C3H5ClMg
Molecular Weight (g/mol) 100.83
MDL Number MFCD00000473
InChI Key PLYLAFITHJUEGX-UHFFFAOYSA-M
SMILES Cl[Mg]CC=C

Specifications

Color Black
CAS Min % 15.76, 80.04
CAS Max % 19.96, 84.25
Linear Formula H2C=CHCH2MgCl
Concentration 1.5 to 1.9M
Density 0.9600g/mL
Synonym allylmagnesium chloride, magnesium, chloro-2-propenyl, magnesium, chloro-2-propen-1-yl, chloro prop-2-en-1-yl magnesium, allylmagnesium chloride solution, 2.0 m in thf, sjpdadpprujp@, allylmagnesiumchloride, allyl magnesium chloride, allyl-magnesium chloride, 2-propenyl magnesium chloride
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name chloro(prop-2-en-1-yl)magnesium
PubChem CID 75804
Formula Weight 100.83
Percent Purity 1.5 to 1.9M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.96
Quantity 800 mL
Flash Point −17°C
Physical Form Liquid
Chemical Name or Material Allylmagnesium chloride, 1.7M Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylmagnesium chloride, 1.7M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'