missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyloxy-tert-butyldimethylsilane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Description
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water (0.50 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water (0.50 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
Boiling Point | 120°C (50mmHg) |
Quantity | 100 mL |
Linear Formula | H2C=CHCH2OSi(CH3)2C(CH3)3 |
UN Number | UN1993 |
Flash Point | 33°C (91°F) |
Solubility Information | Sparingly soluble in water (0.50g/L at 25°C). |
Formula Weight | 172.34 |
Percent Purity | 98% |
Refractive Index | 1.421 |
Density | 0.811 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'