missing translation for 'onlineSavingsMsg'
Learn More
Allylthiourea, 98%, Thermo Scientific Chemicals Product Code.: 10049470

Allylthiourea, 98%, Thermo Scientific Chemicals

Product Code. 10049470
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 10049470

Brand: Thermo Scientific Acros 148801000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-57-9
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70.0°C to 78.0°C
Color White to Beige
Loss on Drying 1.5% max. (50°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H8N2S
Linear Formula H2C=CHCH2NHCSNH2
Quantity 100 g
Beilstein 04, 211
Merck Index 15, 9515
Solubility Information Solubility in water: 67g/L (20°C). Other solubilities: 1370g/L methanol (20°C)- insoluble in benzene,soluble in ether,solutions of borax,benzoates,and urethane,slightly soluble in 70% alcohol
Molecular Weight (g/mol) 116.18
Formula Weight 116.18
Percent Purity 98%
Physical Form Crystals, Powder, Granules or Chunks
Chemical Name or Material Allylthiourea, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465175 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number148801000,148800250,148805000
Lot NumberA0458114 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number148801000,148800250,148805000
Lot NumberA0435149 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number148801000,148800250,148805000
Lot NumberA0403478 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number148801000,148800250,148805000
Lot NumberA0414194 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number148801000,148800250,148805000
compliance-icons
Product Identifier
  • Allylthiourea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allylthiourea, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.