missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Alnico V Magnetic Stirring Bars

Brand:  IKA™

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15340325

  • £14.80 / Pack of 5
Estimated Shipment: 05-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

8 mm
Alnico Magnet
25 mm
5/Pk.
Useful for mixing reagents which resist dissolving or for avoiding any residues at the bottom of the vessels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers