missing translation for 'onlineSavingsMsg'
Learn More

Aloin, Thermo Scientific Chemicals

Aloe exudant, used as a stimulant-laxative

£125.00 - £231.00

Chemical Identifiers

CAS 1415-73-2
Molecular Formula C21H22O9
Molecular Weight (g/mol) 418.40
MDL Number MFCD00151160
InChI Key AFHJQYHRLPMKHU-WEZNYRQKSA-N
Synonym barbaloin, aloin, aloin a, aloinum, aloin ban, dsstox_cid_25967, 10s-1,8-dihydroxy-3-hydroxymethyl-10-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl-10h-anthracen-9-one, 1s-1,5-anhydro-1-9s-4,5-dihydroxy-2-hydroxymethyl-10-oxo-9,10-dihydroanthracen-9-yl-d-glucitol, 10-1',5'-anhydroglucosyl aloe-emodin-9-anthrone, barbaloin a
PubChem CID 12305761
ChEBI CHEBI:2991
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424579
View Documents
Thermo Scientific Alfa Aesar
J62153.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15434579
View Documents
Thermo Scientific Alfa Aesar
J62153.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Aloe exudant, used as a stimulant-laxative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1415-73-2
418.40
AFHJQYHRLPMKHU-WEZNYRQKSA-N
12305761
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12
C21H22O9
MFCD00151160
barbaloin, aloin, aloin a, aloinum, aloin ban, dsstox_cid_25967, 10s-1,8-dihydroxy-3-hydroxymethyl-10-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl-10h-anthracen-9-one, 1s-1,5-anhydro-1-9s-4,5-dihydroxy-2-hydroxymethyl-10-oxo-9,10-dihydroanthracen-9-yl-d-glucitol, 10-1',5'-anhydroglucosyl aloe-emodin-9-anthrone, barbaloin a
CHEBI:2991

Specifications

1415-73-2
C21H22O9
6077558
AFHJQYHRLPMKHU-WEZNYRQKSA-N
(10S)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-10H-anthracen-9-one
12305761
418.4
Aloin
Yellow to Brown
MFCD00151160
barbaloin, aloin, aloin a, aloinum, aloin ban, dsstox_cid_25967, 10s-1,8-dihydroxy-3-hydroxymethyl-10-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl-10h-anthracen-9-one, 1s-1,5-anhydro-1-9s-4,5-dihydroxy-2-hydroxymethyl-10-oxo-9,10-dihydroanthracen-9-yl-d-glucitol, 10-1',5'-anhydroglucosyl aloe-emodin-9-anthrone, barbaloin a
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12
418.40
CHEBI:2991
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'rtecsNumber' : LZ6520000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.