missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Alpelisib, 95%
Brand: Thermo Scientific™ 468121000
557.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Alpelisib | |
(S)-N1-(4-Methyl-5-(2-(1,1,1-trifluoro-2-methylpropan-2-yl)pyridin-4-yl)thiazol-2-yl)pyrrolidine-1,2-dicarboxamide; BYL-719 | |
C19 H22 F3 N5 O2 S | |
Solid | |
95% |
1217486-61-7 | |
100 mg | |
441.47 | |
Reagent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
344412 | Certificate of Analysis | 09/10/2020 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Alpelisib, 95% > 100 mg
Spot an opportunity for improvement?Share a Content Correction