missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-1-Antitrypsin Mouse anti-Human, Rat, Clone: 1A9G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66135-1-IG-20UL
This item is not returnable.
View return policy
Description
The serum glycoprotein a1-AT, is a typical member of the serpin family (serine protease inhibitors), and, an inhibitor of trypsin. a1- AT is synthesized in the liver thus as a single polypeptide chain of approximately 51 kDa showing a marked degree of genetic polymorphism due to the presence of over 20 different alleles. Individuals homozygous for a1-AT defiiency are predisposed to pulmonary emphysema and hepatic cirrhosis.Specifications
Alpha-1-Antitrypsin | |
Monoclonal | |
0.85 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01009, P17475 | |
SERPINA1 | |
Alpha-1-Antitrypsin Fusion Protein Ag9516 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
1A9G6 | |
Unconjugated | |
SERPINA1 | |
A1A, A1AT, AAT, Alpha 1 antiproteinase, Alpha 1 antitrypsin, Alpha 1 protease inhibitor, PI, PI1, PRO2209, PRO2275, Serpin A1, SERPINA1, SPAAT, alpha1-antitrypsin | |
Mouse | |
Protein G | |
RUO | |
24648, 5265 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction