missing translation for 'onlineSavingsMsg'
Learn More

Alpha-1-Antitrypsin Mouse, Clone: AAT/1378, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 5265-MSM1-P1

 View more versions of this product

Product Code. 15960638

  • £395.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Alpha-1-Antitrypsin
Cytoplasmic
AAT/1378
Unconjugated
NP_001121173
Mouse
54kDa
Protein A/G Purified
RUO
5265
IgG1
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
SERPINA1
Recombinant human Alpha-1-Antitrypsin (AAT) protein fragment
HepG2 Cells. Tonsil, Hepatocellular Carcinoma or Histiocytoma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers