missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Actinin 2 Monoclonal Antibody (GT1253), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (94%), Pig (99%), Chicken (98%), Bovine (99%). Positive Control: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it i involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments.Specifications
alpha Actinin 2 | |
Monoclonal | |
1.27 mg/mL | |
PBS with no preservative | |
P35609, Q9JI91 | |
Actn2 | |
Recombinant protein encompassing a sequence within the center region of human alpha Actinin 2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
GT1253 | |
Unconjugated | |
Actn2 | |
1110008F24Rik; actinin alpha 2; actinin, alpha 2; ACTN2; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin-2; CMD1AA; CMH23; F-actin cross-linking protein | |
Mouse | |
Protein G | |
RUO | |
11472, 88 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction