missing translation for 'onlineSavingsMsg'
Learn More
alpha,alpha,alpha-Trifluorotoluene, 99+%, Thermo Scientific Chemicals Product Code.: 10583301

alpha,alpha,alpha-Trifluorotoluene, 99+%, Thermo Scientific Chemicals

Product Code. 10583301
500 mL, Plastic bottle
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10583301

Brand: Thermo Scientific Acros 139775000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-08-8
Molecular Formula C7H5F3
Molecular Weight (g/mol) 146.11
MDL Number MFCD00000372
InChI Key GETTZEONDQJALK-UHFFFAOYSA-N
Synonym benzotrifluoride, trifluoromethyl benzene, alpha,alpha,alpha-trifluorotoluene, phenylfluoroform, benzene, trifluoromethyl, benzenyl fluoride, benzylidyne fluoride, usaf ma-16, a,a,a-trifluorotoluene, trifluoro toluene
PubChem CID 7368
ChEBI CHEBI:36810
IUPAC Name trifluoromethylbenzene
SMILES C1=CC=C(C=C1)C(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -29°C
Color Colorless
Density 1.1900g/mL
Boiling Point 102°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CF3
Refractive Index 1.4130 to 1.415
Quantity 500 mL
Beilstein 05, 290
Merck Index 15, 1113
Specific Gravity 1.19
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in many organic solvents
Formula Weight 146.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluorotoluene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a,a,a-Trifluorotoluene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
alpha,alpha,alpha-Trifluorotoluene, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.