missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dibromo-m-xylene, 97%, Thermo Scientific Chemicals

Product Code. 11741371
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-15-3
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.95
InChI Key OXHOPZLBSSTTBU-UHFFFAOYSA-N
Synonym 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene
PubChem CID 69373
IUPAC Name 1,3-bis(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)CBr)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 75.0°C to 77.0°C
Color White to Brown
Boiling Point 135.0°C to 140.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H4(CH2Br)2
Quantity 5 g
Beilstein 05,374
Solubility Information Solubility in water: insoluble
Formula Weight 263.95
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material α, α'-Dibromo-m-xylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • alpha, alpha'-Dibromo-m-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'