missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Apo-oxytetracycline, 'can be used as secondary standard'

Product Code. 10073270 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 10073270

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alpha-Apo-oxytetracycline, CAS # 18695-01-7, is considered a metabolite formed from oxytetracycline, a member of the tetracycline family, and displays antibiotic activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information
• Alpha-Apo-oxytetracycline is a degradation product of oxytetracycline. When this degradation occurs, two isomers are generated, β- and α -apo-oxytetracycline


Applications
• Alpha-Apo-oxytetracycline compound presents antibiotic activity
• It promotes the inhibition of the growth of aerobic sludge bacteria, Pseudomonas, Agrobacterium, Moraxella, Bacillus, as well as tetracycline-resistant strains of E. coli

TRUSTED_SUSTAINABILITY

Specifications

CAS 18695-01-7
Melting Point 200.0°C
Color Khaki
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Molecular Formula C22H22N2O8
MDL Number MFCD00062827
Quantity 100 mg
Merck Index 10, 6846
Specific Rotation − 64.00
Synonym beta-apo-oxytetracycline, alpha-apo-oxytetracycline, alpha-apooxytetracycline, alpha-apoterramycin, beta-apo oxytetracycline, 2z-2-amino hydroxy methylidene-5-4,5-dihydroxy-9-methyl-3-oxo-1h-benzo f 2 benzofuran-1-yl-4-dimethylamino-6-hydroxycyclohexane-1,3-dione, 4-1,3-dihydro-4,5-dihydroxy-9-methyl-3-oxonaphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxo-1-cyclohexene-1-carboxamide, 4-4,5-dihydroxy-9-methyl-3-oxo-1,3-dihydronaphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide, 4-4,5-dihydroxy-9-methyl-3-oxohydrobenzo f isobenzofuranyl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-enecarboxamide, 4-4,5-dihydroxy-9-methyl-3-oxo-1h-naphtho 2,3-c furan-1-yl-3-dimethylamino-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution and ammonia
InChI Key DRKMHDAKULCOKQ-UHFFFAOYSA-N
SMILES CC1=C2C(OC(=O)C2=C(C3=C1C=CC=C3O)O)C4C(C(=C(C(=O)C4O)C(=O)N)O)N(C)C
IUPAC Name 4-(4,5-dihydroxy-9-methyl-3-oxo-1H-benzo[f][2]benzofuran-1-yl)-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohexene-1-carboxamide
Molecular Weight (g/mol) 442.39
PubChem CID 54710410
Formula Weight 442.39
Percent Purity ≥97.0% (on anhydrous substance), ≥80 wt% (as is, calculated)(HPLC)
Physical Form Crystalline Powder
Chemical Name or Material α-Apo-oxytetracycline, (can be used as secondary standard)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • alfa-Apo-oxytetracycline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.