missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha/beta Tubulin Monoclonal Antibody (TU-10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119400
This item is not returnable.
View return policy
Description
Western Blot: Reducing conditions.
Tubulin protein is the major constituent of microtubules. Tubulin is a dimer of alpha and beta chains, which binds two molecules of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain. There are at least six isotypes of both alpha- and beta- tubulin in human cells, which are distinguished by slightly different amino acid sequences and encoded by a large, multigene family that has been highly conserved throughout evolution. Although the most important functions of microtubules in proliferative cells are through their actions as components of the mitotic spindle, they are also involved in many other essential functions throughout the cell cycle of both malignant and nonmalignant cells. Anti-microtubule agents including Vinca alkaloids and taxanes may disrupt many of these essential functions.Specifications
alpha/beta Tubulin | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
A2AQ07, P02550, P07437, P68369, P68370, Q71U36, Q767L7, Q9H4B7 | |
TUBA1A, TUBB, TUBB1 | |
Microtubule proteins from porcine brain. | |
100 μg | |
Primary | |
Human, Multi-species, Mouse, Porcine, Rat | |
Antibody | |
IgM |
Flow Cytometry, Immunocytochemistry, Western Blot | |
TU-10 | |
Unconjugated | |
TUBA1A | |
2810484G07Rik; Alpha-tubulin 1; Alpha-tubulin 3; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; beta 5-tubulin; beta Ib tubulin; CDCBM6; CSCSC1; Detyrosinated tubulin alpha-1A chain; FLJ30169; H2 ALPHA; hum-a-tub1; hum-a-tub2; LIS3; M(beta)1; M40; OK/SW-cl.56; Talpha1 alpha-tubulin; Tuba1; Tuba-1; TUBA1A; TUBA3; TUBB; TUBB1; TUBB2A; TUBB5; Tubulin alpha; tubulin alpha 1a; tubulin alpha 3; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin beta; tubulin beta 1 class VI; tubulin beta chain; tubulin beta class I; tubulin beta-1 chain; Tubulin beta-5 chain; tubulin, alpha 1; tubulin, alpha 1A; tubulin, alpha, brain-specific; tubulin, beta 1 class VI; tubulin, beta 2A class IIa; tubulin, beta class I; tubulin, beta family 1; tubulin, beta polypeptide; a/β-tubulin | |
Mouse | |
purified | |
RUO | |
100134983, 100135051, 203068, 22142, 545486, 64158, 679312, 733686, 7846, 81027 | |
4° C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction