Learn More
Invitrogen™ Alpha-Bungarotoxin Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Flour and Alexa Flour Plus conjugates.
Brand: Invitrogen™ T1175
452.96 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions. Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor.Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

Specifications
Yellow | |
Tetramethylrhodamine α-Bungarotoxin | |
Store in freezer (–5° to –30°C) and protect from light. | |
Room Temperature | |
Receptors |
Fluorescence | |
500 μg | |
Tetramethylrhodamine | |
Fluorescence Microscope | |
554/577 nm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.