Learn More
Invitrogen™ Alpha-Bungarotoxin Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Flour and Alexa Flour Plus conjugates.
Brand: Invitrogen™ B35450
Code : 50
Additional Details : Weight : 0.00450kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions. Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor.Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).
Specifications
Conjugate | |
Store in freezer (–5° to –30°C) and protect from light. | |
Room Temperature | |
660/668 nm |
500 μg | |
Alexa Fluor™ 647 | |
Other Proteins | |
Alexa Fluor™ |