missing translation for 'onlineSavingsMsg'
Learn More
alpha-Chlorocinnamaldehyde, 90+%, Thermo Scientific™ Product Code.: 10495173

alpha-Chlorocinnamaldehyde, 90+%, Thermo Scientific™

Product Code. 10495173
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10495173

Brand: Thermo Scientific Acros 393530050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18365-42-9
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.61
MDL Number MFCD00077440
InChI Key SARRRAKOHPKFBW-TWGQIWQCSA-N
Synonym alpha-chlorocinnamaldehyde, 2-chloro-3-phenylacrylaldehyde, 2-propenal, 2-chloro-3-phenyl, 2-chloro-3-phenyl-2-propenal, cinnamaldehyde, alpha-chloro, .alpha.-chlorocinnamaldehyde, 2z-2-chloro-3-phenylprop-2-enal, z-2-chloro-3-phenyl-2-propenal, a-chlorocinnamaldehyde, cinnamaldehyde, alpha-chloro-, zShow MoreShow Less
PubChem CID 5899053
IUPAC Name (Z)-2-chloro-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=C(C=O)Cl

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 20°C to 22°C
Color Brown
Boiling Point 107°C to 108°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=C(Cl)CHO
Refractive Index 1.6390 to 1.641
Quantity 5 g
Formula Weight 166.61
Percent Purity 90+%
Physical Form Liquid After Melting
Chemical Name or Material α-chlorocinnamaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • a-Chlorocinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
alpha-Chlorocinnamaldehyde, 90+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.