missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Galactose pentaacetate, Thermo Scientific Chemicals

Product Code. 15417699
Click to view available options
Quantity:
5 g
25 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4163-59-1
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-galactose pentaacetate, a-d-galactose pentaacetate, pentaacetyl-alpha-d-galactose, alpha-d-galactose penta-acetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactopyranose, alpha-d-galactose-pentaacetate, 1,2,3,4,6-pentaacetyl-alpha-d-galactose, alpha-d-galactose 1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-alpha-d-galactose, penta-o-acetyl-alpha-d-galactose
PubChem CID 6432426
ChEBI CHEBI:63145
IUPAC Name [(2R,3S,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 143°C to 144°C
Color White
Quantity 50 g
Solubility Information Soluble in methanol at approximately 50mg/ml
Formula Weight 390.34
Physical Form Powder
Chemical Name or Material alpha-D-Galactose pentaacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'