Learn More
Alpha-Estradiol, MedChemExpress
MedChemExpress Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.
£68.97 - £114.95
Specifications
Chemical Name or Material | Alpha-Estradiol |
---|---|
CAS | 57-91-0 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C18H24O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498126
|
Medchem Express
HY-B0141A |
100 mg |
£68.97
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16412046
|
Medchem Express
HY-B0141A |
500 mg |
£114.95
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alpha-Estradiol is a weak estrogen and a 5α-reductase inhibitor which is used as a topical medication in the treatment of androgenic alopecia.Specifications
Alpha-Estradiol | |
White | |
C18H24O2 | |
DMSO : 62.5 mg/mL (229.46 mM; ultrasonic and warming and heat to 60°C) | |
272.38 | |
99.52% | |
Powder |
57-91-0 | |
Research | |
Alfatradiol Epiestradiol Epiestrol | |
C[C@@]12[C@H](O)CC[C@@]1([H])[C@]3([H])CCC4=C(C=CC(O)=C4)[C@@]3([H])CC2 | |
272.38 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H341∣H351∣H362∣H410
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.