missing translation for 'onlineSavingsMsg'
Learn More

alpha-(Phenylthio)phenylacetic acid, 99%, Thermo Scientific™

Product Code. 11302947
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10490-07-0
Molecular Formula C14H12O2S
Molecular Weight (g/mol) 244.308
MDL Number MFCD00066259
InChI Key UOCQATUFLPHRNG-UHFFFAOYSA-N
Synonym phenyl phenylsulfanyl acetic acid, 2-phenyl-2-phenylthio acetic acid, alpha-phenylthio phenylacetic acid, 2-phenyl-2-phenylsulfanyl acetic acid, 2-phenyl-2-phenylthioacetic acid, acmc-20alu8, phenylthio phenylacetic acid, phenylthio-phenyl acetic acid, phenyl phenylthio acetic acid, alpha-phenylthio-phenylacetic acid
PubChem CID 239605
IUPAC Name 2-phenyl-2-phenylsulfanylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)SC2=CC=CC=C2

Specifications

Melting Point 99°C to 101°C
Quantity 25 g
Beilstein 2373923
Formula Weight 244.31
Percent Purity 99%
Chemical Name or Material alpha-(Phenylthio)phenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'